Order Entry
Spain
ContactUsLinkComponent
SARS-CoV-2 (NS8) Peptide Pool
SARS-CoV-2 (NS8) Peptide Pool
Catalog # STMC100-0663
CAS Number:  
undefined
SARS-CoV-2 (NS8) Peptide Pool
Catalog # STMC100-0663
CAS Number:  

Specifications

  • Env:
    1 KIT
  • Nombre de proteína / péptido:
    SARS-CoV-2 (NS8) Peptide Pool
  • Pureza:
    Average 70%

Specifications

About this item

The SARS-CoV-2 (NS8) Peptide pool is a lyophilised mixture of 28 peptides from non-structural protein 8 (NS8) of SARS-CoV-2. NS8 has been identified as one of the proteins exclusively conserved in a few species of the coronavirus family, and it contains Class II MHC epitopes. The pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 121 on NS8.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').