Order Entry
Spain
ContactUsLinkComponent
Anti-NOTCH3 Rabbit Polyclonal Antibody
Anti-NOTCH3 Rabbit Polyclonal Antibody
Catalog # PRSI15-041
Supplier:  ProSci Inc.
New Product
Anti-NOTCH3 Rabbit Polyclonal Antibody
Catalog # PRSI15-041
Supplier:  ProSci Inc.
New Product

Specifications

  • Tipo de anticuerpo:
    Primario
  • Nombre del antígeno:
    Notch 3
  • Simbolo antígeno:
    NOTCH3
  • Clonalidad:
    Polyclonal
  • Conjugado:
    Unconjugated
  • Huésped:
    Rabbit
  • Isotipo:
    IgG
  • Reactividad:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Forma:
    Liquid
  • ID del gen:
    4854
  • Sinónimos de antígenos:
    Notch (Drosophila) homolog 3|Notch homolog 3 (Drosophila)|HGNC:7883|IMF2|NOTCH3|CASIL|CADASIL|Notch homolog 3
  • Modificación:
    None
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Temperatura de almacenamiento:
    Store at −20 °C. Avoid freeze/thaw cycles
  • Concentración:
    Batch dependent
  • Shipping temperature:
    20
  • Inmunógeno:
    A synthetic peptide corresponding to a sequence within amino acids 2200 to the C-terminus of human NOTCH3 (NP_000426.2)
  • Purification:
    Affinity purification
  • Tipo de env.:
    Vial
  • Env:
    50 µl

Specifications

About this item

Type: Primary
Antigen: NOTCH3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat