Order Entry
Spain
ContactUsLinkComponent
Anti-HNMT Rabbit Polyclonal Antibody
Anti-HNMT Rabbit Polyclonal Antibody
Catalog # PRSI15-026
Supplier:  ProSci Inc.
New Product
Anti-HNMT Rabbit Polyclonal Antibody
Catalog # PRSI15-026
Supplier:  ProSci Inc.
New Product

Specifications

  • Tipo de anticuerpo:
    Primario
  • Nombre del antígeno:
    Histamine N-methyltransferase
  • Simbolo antígeno:
    HNMT
  • Clonalidad:
    Polyclonal
  • Conjugado:
    Unconjugated
  • Huésped:
    Rabbit
  • InmunoQuímica:
    Yes
  • Precipitación inmunológica:
    Yes
  • Isotipo:
    IgG
  • Reactividad:
    Human,
    Mouse
  • Western blot:
    Yes
  • Forma:
    Liquid
  • ID del gen:
    3176
  • Sinónimos de antígenos:
    HNMT|Histamine N-methyltransferase|HMT
  • Modificación:
    None
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Temperatura de almacenamiento:
    Store at −20 °C. Avoid freeze/thaw cycles
  • Concentración:
    Batch dependent
  • Shipping temperature:
    20
  • Inmunógeno:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1-292 of human HNMT (NP_008826.1)
  • Purification:
    Affinity purification
  • Tipo de env.:
    Vial
  • Env:
    50 µl

Specifications

About this item

Type: Primary
Antigen: HNMT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse