Anti-beta-Trcp Rabbit Monoclonal Antibody [clone: D13F10]
Catalog # CSTA4394S
Supplier: Cell Signaling
Specifications
- Tipo de anticuerpo:Primario
- Simbolo antígeno:beta-Trcp
- Clonalidad:Monoclonal
- Clon:D13F10
- Huésped:Rabbit
- Precipitación inmunológica:Yes
- Isotipo:IgG
- Reactividad:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:CNNGEPPRKIIPEKNSLRQT
- Temperatura de almacenamiento:−20 °C
- Shipping temperature:Ambient
- Env:100 µl
Specifications
About this item
Type: Primary
Antigen: beta-Trcp
Clonality: Monoclonal
Clone: D13F10
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey