Anti-STAT3 (Tyr705) Phosphate Rabbit Monoclonal Antibody [clone: D3A7] (Biotinylated)
Catalog # CSTA4093S
Supplier: Cell Signaling
Specifications
- Tipo de anticuerpo:Primario
- Simbolo antígeno:STAT3 (Tyr705) phosphate
- Clonalidad:Monoclonal
- Clon:D3A7
- Conjugado:Biotin
- Huésped:Rabbit
- Isotipo:IgG
- Reactividad:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:ADPGSAAPyLKTKFIC
- Temperatura de almacenamiento:−20 °C
- Shipping temperature:Cold
- Env:100 µl
Specifications
About this item
Type: Primary
Antigen: STAT3 (Tyr705) phosphate
Clonality: Monoclonal
Clone: D3A7
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey