Order Entry
Spain
ContactUsLinkComponent
Anti-Tuberin/TSC2 Rabbit Monoclonal Antibody [clone: D93F12]
Anti-Tuberin/TSC2 Rabbit Monoclonal Antibody [clone: D93F12]
Catalog # CSTA4308S
Supplier:  Cell Signaling
Anti-Tuberin/TSC2 Rabbit Monoclonal Antibody [clone: D93F12]
Catalog # CSTA4308S
Supplier:  Cell Signaling

Specifications

  • Tipo de anticuerpo:
    Primario
  • Simbolo antígeno:
    Tuberin/TSC2
  • Clonalidad:
    Monoclonal
  • Clon:
    D93F12
  • Citometría de flujo:
    Yes
  • Huésped:
    Rabbit
  • FluorescenciaInmunológica:
    Yes
  • Precipitación inmunológica:
    Yes
  • Isotipo:
    IgG
  • Reactividad:
    Hamster,
    Monkey,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Seq:
    02
  • Sequence:
    CQRKRLISSVEDFTEFV
  • Temperatura de almacenamiento:
    –20 °C
  • Shipping temperature:
    Ambient
  • Env:
    100 µl

Specifications

About this item

Type: Primary
Antigen: Tuberin/TSC2
Clonality: Monoclonal
Clone: D93F12
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Hamster;Monkey