Anti-Myc Tag Mouse Monoclonal Antibody [clone: 9B11] (HRP)
Catalog # CSTA2040S
Supplier: Cell Signaling
Specifications
- Tipo de anticuerpo:Primario
- Simbolo antígeno:Myc Tag
- Clonalidad:Monoclonal
- Clon:9B11
- Conjugado:HRP (Horseradish Peroxidase)
- Huésped:Mouse
- Isotipo:IgG2a
- Reactividad:Multiple
- Western blot:Yes
- Temperatura de almacenamiento:–20 °C
- Shipping temperature:Cold
- Env:100 µl
Specifications
About this item
Type: Primary
Antigen: Myc Tag
Clonality: Monoclonal
Clone: 9B11
Conjugation: HRP
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Any species