Anti-GFP jellyfish Rabbit Monoclonal Antibody [clone: D5.1] (Biotinylated)
CSTA2912S
- Tipo de anticuerpo:Primario
- Simbolo antígeno:GFP jellyfish
- Clonalidad:Monoclonal
- Clon:D5.1
- Conjugado:Biotin
- Huésped:Rabbit
- Isotipo:IgG
- Reactividad:Multiple
- Western blot:Yes
- Sequence:CSKGEELFTGVVPILVELD
- Temperatura de almacenamiento:–20 °C
- Shipping temperature:Cold
- Env:100 µl
Type: Primary
Antigen: GFP jellyfish
Clonality: Monoclonal
Clone: D5.1
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Any species