Anti-E-Cadherin Rabbit Monoclonal Antibody [clone: 24E10] (Alexa Fluor 594)
CSTA7687S
- Tipo de anticuerpo:Primario
- Simbolo antígeno:E-Cadherin
- Clonalidad:Monoclonal
- Clon:24E10
- Conjugado:Alexa Fluor® 594
- Huésped:Rabbit
- FluorescenciaInmunológica:Yes
- Isotipo:IgG
- Reactividad:Human,Mouse
- Sequence:RGLDARPEVTRNDVAPTC
- Temperatura de almacenamiento:+4 °C
- Shipping temperature:Cold
- Env:100 µl
Type: Primary
Antigen: E-Cadherin
Clonality: Monoclonal
Clone: 24E10
Conjugation: Alexa Fluor 594
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse