Anti-GFAT1 Rabbit Monoclonal Antibody [clone: D12F4]
Catalog # CSTA5322S
Supplier: Cell Signaling
Specifications
- Tipo de anticuerpo:Primario
- Simbolo antígeno:GFAT1
- Clonalidad:Monoclonal
- Clon:D12F4
- Huésped:Rabbit
- Precipitación inmunológica:Yes
- Isotipo:IgG
- Reactividad:Human,Rat
- Western blot:Yes
- Sequence:CATHGEPSPVNSHPQRSDKNNE
- Temperatura de almacenamiento:–20 °C
- Shipping temperature:Ambient
- Env:100 µl
Specifications
About this item
Type: Primary
Antigen: GFAT1
Clonality: Monoclonal
Clone: D12F4
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat