Anti-Cyclin D1 (Thr286) phosphate Rabbit Monoclonal Antibody [clone: D29B3]
Catalog # CSTA3300S
Supplier: Cell Signaling
Specifications
- Tipo de anticuerpo:Primario
- Simbolo antígeno:Cyclin D1 (Thr286) phosphate
- Clonalidad:Monoclonal
- Clon:D29B3
- Citometría de flujo:Yes
- Huésped:Rabbit
- FluorescenciaInmunológica:Yes
- Precipitación inmunológica:Yes
- Isotipo:IgG
- Reactividad:Monkey,Human
- Western blot:Yes
- Seq:02
- Sequence:CEEEVDLACtPTDVR
- Temperatura de almacenamiento:–20 °C
- Shipping temperature:Cold
- Env:100 µl
Specifications
About this item
Type: Primary
Antigen: Cyclin D1 (Thr286) phosphate
Clonality: Monoclonal
Clone: D29B3
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey