Anti-14-3-3 zeta/delta Rabbit Monoclonal Antibody [clone: D7H5]
Catalog # CSTA7413S
Supplier: Cell Signaling
Specifications
- Tipo de anticuerpo:Primario
- Simbolo antígeno:14-3-3 zeta/delta
- Clonalidad:Monoclonal
- Clon:D7H5
- Huésped:Rabbit
- Isotipo:IgG
- Reactividad:Pig,Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:CKTEGAEKKQQMAREYREKIETELRD
- Temperatura de almacenamiento:−20 °C
- Shipping temperature:Ambient
- Tamaño:100 µl
- Env:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: 14-3-3 zeta/delta
Clonality: Monoclonal
Clone: D7H5
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat,Monkey,Pig