Anti-TAK1 Rabbit Monoclonal Antibody [clone: D94D7]
Catalog # CSTA5206S
Supplier: Cell Signaling
Specifications
- Tipo de anticuerpo:Primario
- Simbolo antígeno:TAK1
- Clonalidad:Monoclonal
- Clon:D94D7
- Huésped:Rabbit
- Precipitación inmunológica:Yes
- Reactividad:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CKKQLEVIRSQQQKRQGTS
- Temperatura de almacenamiento:−20 °C
- Shipping temperature:Ambient
- Env:100 µl
Specifications
About this item
Type: Primary
Antigen: TAK1
Clonality: Monoclonal
Clone: D94D7
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat