Anti-TFIIB Mouse Monoclonal Antibody [clone: 2F6A3H4]
CSTA4169S
- Tipo de anticuerpo:Primario
- Simbolo antígeno:TFIIB
- Clonalidad:Monoclonal
- Clon:2F6A3H4
- Huésped:Mouse
- Precipitación inmunológica:Yes
- Isotipo:IgG2b
- Reactividad:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:Recombinant human TFIIB protein expressed in E.coli.
- Temperatura de almacenamiento:−20 °C
- Shipping temperature:Ambient
- Env:100 µl
Type: Primary
Antigen: TFIIB
Clonality: Monoclonal
Clone: 2F6A3H4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human;Mouse;Rat;Monkey