Anti-ARF1 Rabbit Monoclonal Antibody [clone: D30A3]
CSTA3806S
- Tipo de anticuerpo:Primario
- Simbolo antígeno:ARF1
- Clonalidad:Monoclonal
- Clon:D30A3
- Huésped:Rabbit
- Isotipo:IgG
- Reactividad:Monkey,Human,Mouse
- Western blot:Yes
- Seq:02
- Sequence:RTLtEPSVDFNHSC
- Temperatura de almacenamiento:−20 °C
- Shipping temperature:Ambient
- Env:100 µl
Type: Primary
Antigen: ARF1
Clonality: Monoclonal
Clone: D30A3
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Monkey