Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
1/16 GC ferrule 0.3 mm ID Agilent style 85% Vespel / 15% Graphite PK10 1 * 10 items
SGEA215163
:
Trajan Scientific and Medical
New Product
1/16 GC ferrule 0.3 mm ID Agilent style 85% Vespel / 15% Graphite PK10 1 * 10 items
SGEA215163
:
Trajan Scientific and Medical
New Product
Frequently Bought Together
VWR®, Screw Neck Vials ND8
: VWR Collection
VWR®, Septa GL 45
: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
: VWR Collection
TLC and GLP plates, silica gel 60, unmodified layers, glass backed
: Merck
VWR®, Short Thread Vials ND9
: VWR Collection
VWR® Air Valve
: VWR Collection
Avantor® ACE® C18, HPLC Columns, 5 µm
: Avantor
HPLC columns, Chromolith®
: Merck
TLC plates, silica gel 60, unmodified and chemically modified layers, aluminium backed
: Merck
VWR®, Crimp Neck Headspace Vials ND20
: VWR Collection
Florisil® 60-100 mesh for chromatography
: VWR Chemicals
VWR®, Screw Neck Vials ND13
: VWR Collection
Avantor® Apollo C18, HPLC Columns
: Avantor
Exhaust filters, V3.0
: S.C.A.T.
EPA screw neck vials, N 24
: MACHEREY-NAGEL
VWR®, Screw Neck Vials ND24
: VWR Collection
VWR®, Snap Cap Vials
: VWR Collection