Order Entry
Spain
ContactUsLinkComponent
Muffle Furnace LT9/13/C550 Tmax1300 with lift door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-3253
Supplier:  NABERTHERM
New Product
Muffle Furnace LT9/13/C550 Tmax1300 with lift door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-3253
Supplier:  NABERTHERM