Order Entry
Spain
ContactUsLinkComponent
Muffle Furnace L9/13/B510 Tmax1300 with flap door protective gas connection and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-3214
Supplier:  NABERTHERM
New Product
Muffle Furnace L9/13/B510 Tmax1300 with flap door protective gas connection and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-3214
Supplier:  NABERTHERM