Order Entry
Spain
ContactUsLinkComponent
Muffle Furnace LT15/11/C550 Tmax1100 with lift door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-2856
Supplier:  NABERTHERM
New Product
Muffle Furnace LT15/11/C550 Tmax1100 with lift door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-2856
Supplier:  NABERTHERM