Order Entry
Spain
ContactUsLinkComponent
Muffle Furnace L9/12/C550 Tmax1200 with flap door and protective gas connection, over-temperature limiter and thermocouple port in t he rear wall 1 * 1 items
Catalog # 455-2778
Supplier:  NABERTHERM
New Product
Muffle Furnace L9/12/C550 Tmax1200 with flap door and protective gas connection, over-temperature limiter and thermocouple port in t he rear wall 1 * 1 items
Catalog # 455-2778
Supplier:  NABERTHERM