Order Entry
Spain
ContactUsLinkComponent
Muffle Furnace L9/11/C550 Tmax1100 with flap door and over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-2735
Supplier:  NABERTHERM
New Product
Muffle Furnace L9/11/C550 Tmax1100 with flap door and over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-2735
Supplier:  NABERTHERM