Order Entry
Spain
ContactUsLinkComponent
Muffle Furnace L3/11/B510 Tmax1100 with flap door protective gas connection and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-2529
Supplier:  NABERTHERM
New Product
Muffle Furnace L3/11/B510 Tmax1100 with flap door protective gas connection and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-2529
Supplier:  NABERTHERM