Order Entry
Spain
ContactUsLinkComponent
Human IL-31 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-AP, recombinant human IL-31 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 1.920 Tests
Catalog # ANTIA270412-2096
Supplier:  ANTIBODIES.COM
New Product
Human IL-31 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-AP, recombinant human IL-31 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 1.920 Tests
Catalog # ANTIA270412-2096
Supplier:  ANTIBODIES.COM