Order Entry
Spain
ContactUsLinkComponent
Rat IL-22 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-AP, recombinant mouse IL-22 ELISA standard, and standard reconstitution buffer. This matched antibody p 1 * 576 Tests
Catalog # ANTIA270455-696
Supplier:  ANTIBODIES.COM
New Product
Rat IL-22 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-AP, recombinant mouse IL-22 ELISA standard, and standard reconstitution buffer. This matched antibody p 1 * 576 Tests
Catalog # ANTIA270455-696
Supplier:  ANTIBODIES.COM