Mouse IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-AP, recombinant mouse IL-17A ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 1.920 Tests
Catalog # ANTIA270441-2096
Supplier: ANTIBODIES.COM