Order Entry
Spain
ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human EBV Induced Gene 2/EBI2 protein to be purified and analysed in vitro. 1 * 10 µG
Catálogo # ANTIA325122-10
Supplier:  ANTIBODIES.COM
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human EBV Induced Gene 2/EBI2 protein to be purified and analysed in vitro. 1 * 10 µG
Catálogo # ANTIA325122-10
Supplier:  ANTIBODIES.COM