Order Entry
Spain
ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Melatonin Receptor 1B/MTNR1B protein to be purified and analysed in vitro. 1 * 100 µG
Catalog # ANTIA325230-100
Supplier:  ANTIBODIES.COM
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Melatonin Receptor 1B/MTNR1B protein to be purified and analysed in vitro. 1 * 100 µG
Catalog # ANTIA325230-100
Supplier:  ANTIBODIES.COM