Order Entry
Spain
ContactUsLinkComponent
A heterodimer of CD11c (Phe20-Pro1107) with a C-terminal 6xHis tag + CD18 (Gln23-Asn700) with a C-terminal human Fc tag. The human F c tag consists of amino acids Glu99-Ala330. 1 * 100 µG
Catalog # ANTIA318010-100
Supplier:  ANTIBODIES.COM
New Product
A heterodimer of CD11c (Phe20-Pro1107) with a C-terminal 6xHis tag + CD18 (Gln23-Asn700) with a C-terminal human Fc tag. The human F c tag consists of amino acids Glu99-Ala330. 1 * 100 µG
Catalog # ANTIA318010-100
Supplier:  ANTIBODIES.COM