Order Entry
Spain
ContactUsLinkComponent
Human IL-17F Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-HRP, recombinant human IL-17F ELISA standard, and standard reconstitution buffer. This matched antib 1 * 1.920 Tests
Catalog # ANTIA325320-2096
Supplier:  ANTIBODIES.COM
New Product
Human IL-17F Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-HRP, recombinant human IL-17F ELISA standard, and standard reconstitution buffer. This matched antib 1 * 1.920 Tests
Catalog # ANTIA325320-2096
Supplier:  ANTIBODIES.COM