Order Entry
Spain
ContactUsLinkComponent
Human IL-17A/F Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection anti body, Streptavidin-HRP, recombinant human IL-17A/F ELISA standard, and standard reconstitution buffer. This matched a 1 * 1.920 Tests
Catalog # ANTIA325368-2096
Supplier:  ANTIBODIES.COM
New Product
Human IL-17A/F Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection anti body, Streptavidin-HRP, recombinant human IL-17A/F ELISA standard, and standard reconstitution buffer. This matched a 1 * 1.920 Tests
Catalog # ANTIA325368-2096
Supplier:  ANTIBODIES.COM