Aniline [CAS:62-53-3] 100 ug/ml in Methanol CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store in a refrigerator and protected from light 1 * 1 mL
Catalog # ACSDP804190
Supplier: CPACHEM
New Product
Aniline [CAS:62-53-3] 100 ug/ml in Methanol CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store in a refrigerator and protected from light 1 * 1 mL
Catalog # ACSDP804190
Supplier: CPACHEM
New Product
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck