Order Entry
Spain
ContactUsLinkComponent
Ashing furnace lv9/11/c550 tmax1100 with flap door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
Catalog # NABELV095K11NOTY
Supplier:  NABERTHERM
New Product
Ashing furnace lv9/11/c550 tmax1100 with flap door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
Catalog # NABELV095K11NOTY
Supplier:  NABERTHERM