Disinfectant dose of peracetic acid hydrogen peroxide, SimpleMix, 473 mL Attached Trigger Spray 1 * 12 Bottle
Catalog # VELTDS200-0616Z02E
Supplier: VELTEK ASSOCIATES
New Product
Disinfectant dose of peracetic acid hydrogen peroxide, SimpleMix, 473 mL Attached Trigger Spray 1 * 12 Bottle
Catalog # VELTDS200-0616Z02E
Supplier: VELTEK ASSOCIATES
New Product
Frequently Bought Together
Acetone, anhydrous (max. 0.01% H₂O) ≥99.8%
Supplier: VWR Chemicals
1,2-Dichlorobenzene, EMSURE® for analysis, Supelco®
Supplier: Merck
Water ≥99.9%, HiPerSolv CHROMANORM®, PFAS grade for LC-MS
Supplier: VWR Chemicals
Benzyl alcohol ≥99.5%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
Benzoic acid methyl ester for synthesis, Sigma-Aldrich®
Supplier: Merck
Sulphuryl chloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Dichloromethane stabilised, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Acetic anhydride for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Water with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
Petroleum spirit, 40…60 °C
Supplier: Thermo Fisher Scientific
Chlorobenzene ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
o-Xylene ≥99%
Supplier: Thermo Fisher Scientific