MAGNESIUM SULFATE ANHYDROUS USP 1 * 12 kg
SPCMM1354-12KG
New Product
Frequently Bought Together
Potassium iodide ≥99%, GPR RECTAPUR®
: VWR Chemicals
Sodium chloride 99.0-100.5% USP, Ph. Eur., BP
: VWR Chemicals
Ammonium iron(II) sulphate hexahydrate ≥98.5%, TECHNICAL
: VWR Chemicals
Sodium salicylate ≥99.5%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Hydrazinium sulphate ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Aluminium potassium sulphate dodecahydrate ≥97%, TECHNICAL
: VWR Chemicals