ZOLMITRIPTAN FOR SYSTEM SUITABILITY CRS 1 * 10 mg
Catalog # EDQMY0001991
Supplier: EDQM
New Product
Frequently Bought Together
VWR®, Osmolality standards
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C
Supplier: REAGECON
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
METALLOID/HYDRIDE ELEMENTS IN 20% HCL, TR. HF 1 * 100 mL
Supplier: LGC Standards PROMOCHEM
Trypsin, Reference Standards
Supplier: USP
VWR®, Cold Cranking Simulator (CCS) viscosity standards
Supplier: VWR Chemicals
Buffer powder pillows
Supplier: Hach
Buffer solutions, colour coded, Fluka™
Supplier: Honeywell Chemicals
VWR®, Cone and Plate viscosity standards
Supplier: VWR Chemicals
Pesticides 2 - WP, proficiency testing material, Supelco®
Supplier: Merck