ZIKA VIRUS -NS1 (SURINAME STRAIN) 1 * 100 µG
Catalog # ENZOENZPRT2750100
Supplier: ENZO LIFE SCIENCES
ZIKA VIRUS -NS1 (SURINAME STRAIN) 1 * 100 µG
Catalog # ENZOENZPRT2750100
Supplier: ENZO LIFE SCIENCES
Frequently Bought Together
Ammonium acetate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Acetic acid 99% ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test strips, nitrite, MQuant®, Supelco®
Supplier: Merck
Test strips for peracetic acid, MQuant®, Supelco®
Supplier: Merck
Cuvettes and tubes for Picco, Multy, Nova and Prove
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Schiff reagent for histology, Sigma-Aldrich®
Supplier: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Test strips for iron, MQuant®, Supelco®
Supplier: Merck
Test strips for ammonium, MQuant®, Supelco®
Supplier: Merck