AMPIVIEW HPV HIGH-RISK RNA PROBES SET 1 * 2 mL
Catalog # ENZOENZGEN1482000
Supplier: ENZO LIFE SCIENCES
AMPIVIEW HPV HIGH-RISK RNA PROBES SET 1 * 2 mL
Catalog # ENZOENZGEN1482000
Supplier: ENZO LIFE SCIENCES
Frequently Bought Together
Laboratory trays
Supplier: KARTELL
VWR® Media Bottles, Square, PETG
Supplier: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
Microcentrifuge tubes, 0,6/1,5/1,7/2,0/5,0 ml, SnapLock, Axygen®
Supplier: Corning
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
PCR plates, 96-well, fully skirted, low profile, Thermo-Fast®
Supplier: Thermo Fisher Scientific
Eppendorf Tubes® Microtubes with Screw Cap, 5,0 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Cell culture storage bottles, square, PC, Corning®
Supplier: Corning
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
Round-bottom polystyrene tubes, disposable, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Cryoboxes, 133×133 mm
Supplier: RATIOLAB
Lab blender bags, Stomacher®
Supplier: Seward (FermionX)