0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Spain
ContactUsLinkComponent
NERATINIB (FREE BASE) 1 * 200 mg
ENZOENZCHM2280200
:
ENZO LIFE SCIENCES
NERATINIB (FREE BASE) 1 * 200 mg
ENZOENZCHM2280200
:
ENZO LIFE SCIENCES
Frequently Bought Together
Visual colour kits, ammonium, MQuant®, Supelco®
: Merck
Test kits, nitrate, Spectroquant®, Supelco®
: Merck
Test kits, surfactants, Spectroquant®, Supelco®
: Merck
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
Test kits, nitrite, Spectroquant®, Supelco®
: Merck
Test kits, phosphate, Spectroquant®, Supelco®
: Merck
Test kits, ammonium, Spectroquant®, Supelco®
: Merck
pH indicator strips, MQuant®, Supelco®
: Merck
Powder pillows, ammonia
: Hach