INFIGRATINIB (FREE BASE) 1 * 25 mg
Catalog # ENZOENZCHM2250025
Supplier: ENZO LIFE SCIENCES
Frequently Bought Together
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Visual colour kits, iron, MQuant®, Supelco®
Supplier: Merck
Buffer tablets according to Weise, Sigma-Aldrich®
Supplier: Merck
Ascorbic acid test strips, Dosatest
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
NANOCOLOR® Photometric Tube Test Kits for Nitrite
Supplier: MACHEREY-NAGEL
VWR®, OCT Embedding matrix for frozen sections
Supplier: VWR Chemicals
Sodium hydroxide ≥98%, pellets, EMPROVE® ESSENTIAL Ph. Eur., BP, FCC, JP, NF, E524, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Chinese ink, green, Q Path® stain for histology
Supplier: VWR Chemicals
Phloxin B for histology, for cytology
Supplier: RAL
Kjeldahl tablets, Kjeltabs
Supplier: THOMPSON & CAPPER
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach