18.2 MO/cm, up to 2.0 l/min Flow rate up to 2.0 l/min Inorganic substances18.2 MO/cm Organic substances (TOC) <5 ppb 1 * 1 items
Catalog # ELGAEPLFLEX03
Proveedor: ELGA LabWater
18.2 MO/cm, up to 2.0 l/min Flow rate up to 2.0 l/min Inorganic substances18.2 MO/cm Organic substances (TOC) <5 ppb 1 * 1 items
Catalog # ELGAEPLFLEX03
Proveedor: ELGA LabWater
Frequently Bought Together
Pipette controller, accu-jet® S
Proveedor: Brand
Accessories for water purification systems, arium®
Proveedor: Sartorius
VWR®, Digital Timer, One Channel
Proveedor: VWR Collection
VWR® TD20, TD21, TD22 Precise, Universal, Waterproof Thermometers
Proveedor: VWR Collection
3-Packs, Research® plus (General Lab Product)
Proveedor: EPPENDORF
VWR®, Digital Timer, One Channel
Proveedor: VWR Collection
VWR®, Digital Timer, One Channel
Proveedor: VWR Collection
VWR®, Temperature Indicating Strips
Proveedor: VWR Collection
tempmate.®-S1 Single Use Temperature Data Logger
Proveedor: tempmate
VWR®, Digital Timer, One Channel
Proveedor: VWR Collection
VWR® Pipette Controller
Proveedor: VWR Collection
VWR®, Digital Timer, Four Channels, with Clock
Proveedor: VWR Collection
VWR®, Digital Timer, One Channel
Proveedor: VWR Collection
Eppendorf® Pipette Holder System
Proveedor: EPPENDORF
Accessories for marco
Proveedor: Brand
Cartridges for Milli-Q®
Proveedor: Merck