Dropping bottle, LDPE
Cap, white, 20 mm
Catalog # HACH2683806
Supplier: Hach
Jump to:
Product Details & Documents
LDPE, translucent.
This dropping bottle is supplied without dropper plug and cap. Plugs and caps must be ordered separately.
Specifications
- Description:Cap, white, 20 mm
- Seq:03