Order Entry
Spain
Orders LinkContactUsLinkComponent
Temperature indicating strips
Temperature indicating strips
121…160 °C
Catalog # SIMETC8030
Supplier:   Industrial Physics
Temperature indicating strips
Catalog # SIMETC8030
Supplier:  Industrial Physics
Temperature indicating strips
Temperature indicating strips
121…160 °C
Price
32,60 €
Log in for availability
1 * 10 UN
Product Details & Documents
These temperature indicating strips are designed for use in electronics, food and pharmaceutical industries, and also in the oven and engine construction businesses. They are particularly useful for indicating temperatures of moving parts.
  • Self adhesive foils to measure maximum surface temperatures
  • Indicators change colour when marked temperature is reached and the change is irreversible
  • Highest temperature remains visible for months
Delivery information: Series 5 and Series 8 contains 10 stickers per sheet.
Specifications
  • Seq:
    11
  • Range:
    121…160 °C
Recomendaciones
Las recomendaciones se personalizarán según sus preferencias de compra solo si ha dado su consentimiento activando la opción "Mejorar mi experiencia de compra" en la "página de información personal".
De lo contrario, recibirá recomendaciones genéricas.