Cytidine 5Æ-triphosphate (CTP) is supplied as a 100 mM aqueous solution at pH 7.4. 1 * 1 mL
ABNOU0436.1ML
: Abnova
Cytidine 5Æ-triphosphate (CTP) is supplied as a 100 mM aqueous solution at pH 7.4. 1 * 1 mL
ABNOU0436.1ML
: Abnova
Frequently Bought Together
VWR®, Spreaders, T-shaped
: VWR Collection
Reagent/centrifuge tube with screw cap
: Greiner Bio-One
Transfer pipettes
: RATIOLAB
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Disposable Microcentrifuge Tubes
: VWR Collection
VWR®, Petri Dish, PS
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection