Order Entry
Spain
ContactUsLinkComponent
Furnace ashing lv9/11/c550 tMax1100 with FlaP Door anD over-teMPerature liMiter anD therMocouPle hole in the Furnace Door 1 * 1 items
Catalog # NABELV095K11NOTZ1
Supplier:  NABERTHERM
Furnace ashing lv9/11/c550 tMax1100 with FlaP Door anD over-teMPerature liMiter anD therMocouPle hole in the Furnace Door 1 * 1 items
Catalog # NABELV095K11NOTZ1
Supplier:  NABERTHERM
Frequently Bought Together