Neutralized sodium hypochlorite 0.25% solution, attached activator, SimpleMix, 1 Gallon 1 * 4 Bottle
VELTSHC-NPH0.25-02
Neutralized sodium hypochlorite 0.25% solution, attached activator, SimpleMix, 1 Gallon 1 * 4 Bottle
VELTSHC-NPH0.25-02
Frequently Bought Together
Acetonitrile, ChromAR® for HPLC, Macron Fine Chemicals™
: MACRON AVANTOR BRAND
3-(Chloromethyl)benzoyl chloride 98%
: Thermo Fisher Scientific
n-Nonane 99%, Reagent Grade
: Honeywell Chemicals
Glycerol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 5 L
: PanReac AppliChem
Methanol ACS, gradient grade for HPLC, Burdick & Jackson™
: Honeywell Chemicals
Ethanol absolute Ph. Eur., J.T.Baker®
: Avantor
4,4'-Bis(dimethylamino)diphenylmethane 98%
: Thermo Fisher Scientific
Bromoform 96% stabilised
: Thermo Fisher Scientific
Isooctane (Reag. Ph. Eur.) for analysis, ACS 1 * 2,5 L
: PanReac AppliChem
Hexane (mixture of isomers), TECHNICAL
: VWR Chemicals