0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Spain
ContactUsLinkComponent
REAGENT KIT FOR PRIMELAB ID85 (50 TESTS) 1 * 1 items
WTIDPL85-KIT
:
WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID85 (50 TESTS) 1 * 1 items
WTIDPL85-KIT
:
WATER ID PRIMELAB
Frequently Bought Together
pH indicator papers, MQuant®, Supelco®
: Merck
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Test kits, nitrate, Spectroquant®, Supelco®
: Merck
May Grunwald's eosin-methylene blue modified solution stain for hematology, for cytology, Sigma-Aldrich®
: Merck
Test strips for peroxide, MQuant®, Supelco®
: Merck
Test kits, surfactants, Spectroquant®, Supelco®
: Merck
Test kits, nitrite, Spectroquant®, Supelco®
: Merck
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
Test Strips, nitrate, MQuant®, Supelco®
: Merck
Test kits, ammonium, Spectroquant®, Supelco®
: Merck
Ethanol 70% ≥70% (v/v)
: VWR Chemicals
Sodium hydroxide, pellets, pure, Supelco®
: Merck
pH indicator strips, MQuant®, Supelco®
: Merck