REAGENT KIT FOR PRIMELAB ID52 (50 TESTS) 1 * 1 items
Catalog # WTIDPL52-KIT
Supplier: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID52 (50 TESTS) 1 * 1 items
Catalog # WTIDPL52-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
UltraClear® for histology, J.T.Baker®
Supplier: Avantor
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
SODIUM HYPOCHLORITE 12,5 0,1 N 1 * 1 l
Supplier: PROLABO CMP
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Formaldehyde 36% in aqueous solution stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Reagents for copper determination using Lovibond® MD photometers
Supplier: Lovibond
J.T.Baker® Endonuclease
Supplier: Avantor
Acid Red 183 30% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
ALKALINE-CYANIDE REAGENT 1 * 500 mL
Supplier: Hach
PHENOLPHTHALEIN 1% W/V IN PYRIDINE 1 * 100 mL
Supplier: CUSTOM MADE CHEMICALS LAB
PHENOLRED PHOTOMETER TABLETS 1 * 500 Tablet
Supplier: Lovibond