REAGENT KIT FOR PRIMELAB ID31 (50 TESTS) 1 * 1 items
Catalog # WTIDPL31-KIT
Supplier: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID31 (50 TESTS) 1 * 1 items
Catalog # WTIDPL31-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Test strips for peracetic acid, MQuant®, Supelco®
Supplier: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test strips, zinc, MQuant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Test strips for free chlorine, MQuant®, Supelco®
Supplier: Merck
Test kits, surfactants, Spectroquant®, Supelco®
Supplier: Merck
Hydrogen peroxide 6% (w/v) (20 volumes), GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips, sulphite, MQuant®, Supelco®
Supplier: Merck
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Ammonia solution 25%, TECHNICAL
Supplier: VWR Chemicals
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck