REAGENT KIT FOR PRIMELAB ID18 (50 TESTS) 1 * 1 items
Catalog # WTIDPL18-KIT
Supplier: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID18 (50 TESTS) 1 * 1 items
Catalog # WTIDPL18-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
6-CFDA (6-Carboxyfluorescein diacetate, single isomer), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
BUFFER AMMONIA (54 G/L 350 ML/L) 1 * 5 l
Supplier: PROLABO CMP
May Grunwald's eosin-methylene blue, pure stain
Supplier: Thermo Fisher Scientific
SODIUIM HYDROXIDE 30% SOLUTION 1 * 500 mL
Supplier: Spectrum Chemical
65ML PL POLYACRYLATE 2 (65 TESTS) 1 * 65 mL
Supplier: WATER ID PRIMELAB
Methylene blue solution R in ethanol 95%
Supplier: SIGMA ALDRICH MICROSCOPY
Buffer concentrate, citrate phosphate, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Sodium hydroxide 98%, flakes, pure
Supplier: Thermo Fisher Scientific