REAGENT KIT FOR PRIMELAB ID162(50 TESTS) 1 * 1 items
Catalog # WTIDPL162-KIT
Supplier: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID162(50 TESTS) 1 * 1 items
Catalog # WTIDPL162-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test strips for peracetic acid, MQuant®, Supelco®
Supplier: Merck
Test strips, peracetic acid, Dosatest
Supplier: VWR Chemicals
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
VWR®, Powder packs for determination of chlorine, reagents for photometers
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
VWR® Rota®, pH Indicator Paper, 1 to 14
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck